Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-G 1005
GREENER_CHOICE

Catalog No. p-7082172
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
512910 10 mg
512950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 512910 Supplier Tocris Bioscience™ Supplier No. 5129/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective GPBA receptor (TGR5) agonist

TC-G 1005 is a potent and selective GPBA agonist (EC50 values are 0.72 nM and 6.2 nM for hTGR5 and mTGR5, respectively). TC-G 1005 is selective for TGR5 over FXR (farnesoid X receptor). TC-G 1005 increases plasma GLP-1 levels and reduces blood glucose in mice. Orally bioavailable.

Specifications

Chemical Name or Material TC-G 1005
CAS 1415407-60-1
Synonym (4-Cyclopropyl-3,4-dihydro-1(2H)-quinoxalinyl)[4-(2,5-dimethylphenoxy)-3-pyridinyl]methanone
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C25H25N3O2
Formula Weight 399.48
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.