Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0422288
GREENER_CHOICESDP

Catalog No. 537850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
537850 50 mg
537810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 537850 Supplier Tocris Bioscience™ Supplier No. 5378/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective group III mGlu positive allosteric modulator

VU 0422288 is a group III mGlu receptor positive allosteric modulator (EC50 values are 108, 125 and 146 nM for mGlu4, mGlu8 and mGlu7 respectively). Exhibits selectivity over a panel of 68 other GPCRs, ion channels and transporters. Potentiates mGlu7-mediated reductions in excitatory post synaptic potentials in hippocampal slices.

Specifications

Chemical Name or Material VU 0422288
CAS 1630936-95-6
Synonym N-[3-Chloro-4-[(5-chloro-2-pyridinyl)oxy]phenyl]-2-pyridinecarboxamide
Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C17H11Cl2N3O2
Formula Weight 360.19
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.