Learn More
Description
- This low molecular weight peptide represents a minimal sequence of C-terminal residues of ubiquitin where hydrolysis occurs at the UbGly76-X bond
- The catalytic efficiency for hydrolysis of this substrate by deconjugating enzymes is several thousand-fold less than a more native substrate as remote interactions with Ub-containing substrates stabilize catalytic transition states
- Purity >95% by HPLC
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Quantity | 5 mg |
| Product Type | Z-LRGG-AMC, C34H44N8O8 |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
