Learn More
Enzo Life Sciences R-(+)-DIOA (25mg). CAS: 81166-47-4
SDP
Supplier:  Enzo Life Sciences ALX550263M025
Encompass_Preferred
Inhibitor of the K+/Cl--cotransport that does not affect the bumetanide-sensitive Na+/K+/Cl--cotransport system. Alternative name: R-(+)-Butylindazone, (Dihydroindenyl)oxy alkanoic acid. Purity: ≥98% (HPLC). Appearance: White solid. Solubility: Soluble in 100% ethanol or dilute aqueous base; insoluble in water or dilute aqueous acid. Long Term Storage: -20°C.
Catalog No. 50-200-9515