Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-DM-SEGPHOS(regR) 95.0+%, TCI America™
SDP

Catalog No. D4499200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4499200MG 200 mg
D44991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4499200MG Supplier TCI America Supplier No. D4499200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 850253-53-1
Molecular Formula C46H44O4P2
Molecular Weight (g/mol) 722.802
InChI Key XJJVPYMFHXMROQ-UHFFFAOYSA-N
Synonym (R)-(+)-5,5′C-Bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (R)-(+)-4,4′C-Bis[di(3,5-xylyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene)
PubChem CID 11297101
IUPAC Name [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 200 mg
Formula Weight 722.80
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-DM-SEGPHOS(regR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.