Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-Glycidol 98.0+%, TCI America™
SDP

Catalog No. G036325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G036325G 25 g
G03635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G036325G Supplier TCI America Supplier No. G036325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57044-25-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00074873
InChI Key CTKINSOISVBQLD-GSVOUGTGSA-N
Synonym (R)-(+)-1-Hydroxy-2,3-epoxypropane, (R)-(+)-2-(Hydroxymethyl)oxirane
PubChem CID 5460455
ChEBI CHEBI:18664
IUPAC Name [(2R)-oxiran-2-yl]methanol
SMILES C1C(O1)CO

Specifications

Color Yellow
Boiling Point 57°C
Quantity 25 g
UN Number 2810
Formula Weight 74.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-Glycidol
EINECSNumber (2)-2389
RTECSNumber RR0508000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.