Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Butyrylcholinesterase/BCHE Quantikine ELISA Kit
SDP

Catalog No. DBCHE0 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DBCHE0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DBCHE0 Supplier R&D Systems™ Supplier No. DBCHE0
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sufficient for at least one 96-well plates

The Human Butyrylcholinesterase/BCHE Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Butyrylcholinesterase/BCHE in cell culture supernates (50 ul), cell lysates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), saliva (50 ul), urine (50 ul), human milk (50 ul).

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.06 ng/mL
Detectable Analytes Butyrylcholinesterase/BCHE
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Cell Lysates, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Tissue Lysates, Urine
Specificity Natural and Recombinant Human BCHE
For Use With (Application) ELISA
Gene ID (Entrez) 590
Gene Symbol BCHE
Host Species Human
Measuring Range 0.156-10ng/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Saliva (50uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym Acylcholine acylhydrolase, BCHE, Butyrylcholine esterase, butyrylcholinesterase, CHE1, CHE1cholinesterase, Choline esterase II, cholinesterase 1, E1, EC 3.1.1.8, Pseudocholinesterase
Target Butyrylcholinesterase/BCHE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.