Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Calbindin D Quantikine ELISA Kit
SDP

Catalog No. DCALD0 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DCALD0 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DCALD0 Supplier R&D Systems™ Supplier No. DCALD0

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sufficient for at least one 96-well plates

The Human Calbindin D Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Calbindin D/S100G in tissue lysates (10 ul), urine (25 ul).

Specifications

Assay Range 0.08 - 5ng/mL
Assay Sensitivity 0.015ng/mL
Detectable Analytes Calbindin D
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Tissue Lysates, Urine
Specificity Natural and Recombinant Human Calbindin D
For Use With (Application) ELISA
Gene ID (Entrez) 795
Host Species Human
Measuring Range 0.078-5ng/mL
Quantity 1 Kit
Sample Volume Tissue Lysates (10uL),Urine (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.