Learn More
Description
Specifications
Specifications
| Assay Range | 125.00 - 8000pg/mL |
| Assay Sensitivity | 13.8pg/mL |
| Detectable Analytes | IGFBP-1 |
| Assay | Solid Phase Sandwich ELISA |
| Conjugate | HRP |
| Cross Reactivity | < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules. |
| Immunoassay Kit Format | 96-well Strip Plate |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Human Milk, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant phosphorylated and non-phosphorylated Human free IGFBP-1. This assay does not detect IGFBP-1 in. complex with IGF-I or IGF-II. |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
