Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human His10-cIAP-1/HIAP-2 Protein, CF

Catalog No. E3280050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3280050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3280050 Supplier R&D Systems™ Supplier No. E3280050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Recombinant Human cIAP-1/HIAP-2 is a RING finger Ubiquitin ligase (E3) that functions downstream of a Ubiquitin-activating (E1) enzyme and a Ubiquitin--conjugating (E2) enzyme to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial cIAP-1 concentration of 0.2--1 μM.

Specifications

Accession Number Q13490
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 8.0, 500 mM NaCl, 20% Glycerol (v/v), 5 mM TCEP, 0.5 mM EDTA
Gene ID (Entrez) 329
Molecular Weight (g/mol) M.W. (theoretical): 72 kDa
Name His10 cIAP-1/HIAP-2
Quantity 50 μg
Source E. coli-derived human cIAP-1/HIAP-2 protein Contains an N-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Without Additives Carrier Protein
Gene Symbol BIRC2
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein cIAP-1/HIAP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.