Learn More
R&D Systems™ Human IL-1 RAcP/IL-1 R3 (NP_002173) VersaClone cDNA
Shop All Novus Biologicals Products
Description
Order Info
Ships at ambient temperature
Specifications
Specifications
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans