Learn More
R&D Systems™ Human KCC1/SLC12A4 (NP_005063) VersaClone cDNA
SDP
Catalog No. RDC1100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1100 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1100 Supplier R&D Systems™ Supplier No. RDC1100
Add to Cart
Add to Cart
This item is not returnable. View return policy
K-Cl Cotransporter 1
The Human KCC1/SLC12A4 (NP_005063) VersaClone cDNA clone contains the complete ORF for KCC1/SLC12A4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The KCC1/SLC12A4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans