Learn More
R&D Systems™ Human LILRB3/CD85a/ILT5 (NP_006855) VersaClone cDNA
Shop All Novus Biologicals Products

Description
Order Info
Ships at ambient temperature
Specifications
Specifications
| Gene | ILT5/CD85a/LILRB3 |
| Accession Number | NP_006855 |
| Gene ID (Entrez) | 11025 |
| Gene Alias | CD85 antigen-like family member A, CD85a, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3, LIR3CD85A, LIR-3 mgC138403, member 3, Monocyte inhibitory receptor HL9, PIRB |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
For Reasearch Use Only; Not for Use in Humans
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.