Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Neuropilin-1 Quantikine ELISA Kit
SDP

Catalog No. DNRP10 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DNRP10 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DNRP10 Supplier R&D Systems™ Supplier No. DNRP10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Neuropilin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Neuropilin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Neuropilin-1 in cell culture supernates (50 ul), cell lysates (13 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul), human milk (10 ul).

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 9.33pg/mL
Detectable Analytes Neuropilin-1
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Cell Lysates, Human Milk, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human Neuropilin-1
For Use With (Application) ELISA
Gene ID (Entrez) 8829
Host Species Human
Measuring Range 78.1-5,000pg/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (13uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL),Human Milk (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym BDCA4, BDCA-4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, Neuropilin1, NP1, NRP, NRP1, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.