Learn More
R&D Systems™ Human SLC4A7 (NP_003606) VersaClone cDNA
SDP
Catalog No. RDC1015 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC1015 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC1015 Supplier R&D Systems™ Supplier No. RDC1015
Add to Cart
Add to Cart
This item is not returnable. View return policy
Solute Carrier Family 4 (Sodium Bicarbonate Cotransporter), Member 7S
The Human SLC4A7 (NP_003606) VersaClone cDNA clone contains the complete ORF for SLC4A7/Sodium bicarbonate cotransporter 3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SLC4A7/Sodium bicarbonate cotransporter 3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans