Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ST2/IL-33R DuoSet ELISA
SDP

Catalog No. DY523B05 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY523B05 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY523B05 Supplier R&D Systems™ Supplier No. DY523B05
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human ST2/IL-33 R DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human ST2/IL-33 R DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human ST2/IL-33R.

Specifications

Assay Sensitivity 0pg/mL
Detectable Analytes ST2/IL-1 R4
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
For Use With (Application) ELISA
Gene ID (Entrez) 9173
Host Species Human
Measuring Range 31.2-2,000pg/mL
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed
Target ST2/IL-33R
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.