Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human t-Plasminogen Activator/tPA Quantikine ELISA Kit
SDP

Catalog No. DTPA00 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DTPA00 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DTPA00 Supplier R&D Systems™ Supplier No. DTPA00
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human t-Plasminogen Activator/tPA Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human t-Plasminogen Activator/tPA Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human t-Plasminogen Activator/tPA in cell culture supernates (50 ul), serum (13 ul), edta plasma (13 ul), heparin plasma (13 ul), citrate plasma (13 ul), saliva (50 ul), urine (25 ul), human milk (13 ul).

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 16.1pg/mL
Detectable Analytes t-Plasminogen Activator/tPA
Assay Solid Phase Sandwich ELISA
Conjugate HRP
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well Strip Plate
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Citrate, Heparin), Saliva, Serum, Urine
Specificity Natural and Recombinant Human tPA. This assay also recognizes tPA complexed with PAI-1.
For Use With (Application) ELISA
Gene ID (Entrez) 5327
Host Species Human
Measuring Range 78.1-5,000pg/mL
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (13uL),EDTA Plasma (13uL),Heparin Plasma (13uL),Citrate Plasma (13uL),Saliva (50uL),Urine (25uL),Human Milk (13uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym Alteplase, DKFZp686I03148, EC 3.4.21, EC 3.4.21.68, plasminogen activator, tissue, plasminogen activator, tissue type, PLAT, reteplase, tissue plasminogen activator (t-PA), tissue-type plasminogen activator, TPA, T-PA, t-plasminogen activator, tPlasminogen Activator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.