Learn More
Strem, An Ascensus Company CAS 164858-78-0. 250mg. (R)-()-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-45-dihydrooxazole 98% (R)-iPr-PHOX. MFCD02684553
SDP
Supplier:  Strem, An Ascensus Company 151821250MG
Encompass_Preferred
CAS 164858-78-0. 250mg. (R)-()-2-[2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-45-dihydrooxazole 98% (R)-iPr-PHOX. MFCD02684553. Molecular Weight 373.44. Molecular Formula C24H24NOP. Color/form white pwdr. Strem 15-1821. http//www.strem.com/catalog/v/15-1821/
Catalog No. 50-211-0756
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.