Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse PIR-B Protein, CF

Catalog No. 2754PB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-54P-B050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-54P-B050 Supplier R&D Systems™ Supplier No. 2754PB050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When biotinylated recombinant rat MAG/Siglec-4a Fc Chimera is catured on Streptavidin coated plate (Catalog # CP003 ), Recombinant Mouse PIR-B binds with an apparent Kd < 100 nM.

Specifications

Accession Number AAG01012
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with trehalose
Gene ID (Entrez) 11025
Molecular Weight (g/mol) M.W. (Observed): 85-95 kDa, reducing conditions; M.W. (theoretical): 69 kDa
Name PIR-B
Preparation Method Reconstitute at 250μg/mL in sterile PBS.
Quality Control Testing SDS-PAGE: 85-95kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse PIR-B protein Gly24-Gly635, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Gly24
Without Additives Carrier Protein
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Pirb
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein PIR-B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.