Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N-Benzyl-1-phenylethylamine, ChiPros 99+%, ee 96+%, Thermo Scientific™

Catalog No. AAL1929814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1929814 25 g
AAL1929806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1929814 Supplier Thermo Scientific Chemicals Supplier No. L1929814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38235-77-7
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD00015010
InChI Key ZYZHMSJNPCYUTB-CYBMUJFWSA-N
Synonym r-+-n-benzyl-1-phenylethylamine, r-n-benzyl-1-phenylethanamine, r-+-n-benzyl-alpha-methylbenzylamine, benzyl 1r-1-phenylethyl amine, r-n-benzyl-1-phenylethylamine, 1r-1-phenylethyl benzylamine, 1r-n-benzyl-1-phenylethanamine, s---n-benzyl-alpha-phenethylamine, pubchem5974
PubChem CID 1268086
IUPAC Name (1R)-N-benzyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NCC2=CC=CC=C2

Specifications

Density 1.01
Boiling Point 171°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 25 g
Beilstein 3589990
Sensitivity Air Sensitive
Optical Rotation +39° (Neat)
Formula Weight 211.31
Percent Purity ≥99%
Assay ee ≥96%
Chemical Name or Material (R)-(+)-N-Benzyl-1-phenylethylamine
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 609-537-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.