Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ProDots Recombinant Human/Mouse/Rat Activin A Protein

Catalog No. PRD33810
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRD33810 50 μg
PRD33850 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRD33810 Supplier R&D Systems™ Supplier No. PRD33810

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Includes

Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human/Mouse/Rat Activin A.

Supplied as ready-to-use 5 ug protein balls

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.

Order Info

Add ProDots proteins directly to the medium or reconstitute in the vial at 100μg/mL in 4mM HCl.

Specifications

Accession Number P08476
For Use With (Application) Bioactivity
Gene ID (Entrez) 3624
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions, 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer)
Name ProDots™ Recombinant Human/Mouse/Rat Activin A
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2°C to 8°C as supplied. 1 month at 2°C to 8°C under sterile conditions after reconstitution. 3 months (at -20°C to -80°C under sterile conditions after reconstitution.)
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol INHBA
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution For a stock solution, reconstitute at 100 μg/mL in 4 mM HCl, or simply roll ProDot™ directly into cell culture medium for immediate use.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Activin A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.