Learn More
R&D Systems™ ProDots Recombinant Human/Mouse/Rat Activin A Protein
Description
Includes
Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human/Mouse/Rat Activin A.
Order Info
Add ProDots proteins directly to the medium or reconstitute in the vial at 100μg/mL in 4mM HCl.
Specifications
Specifications
| Accession Number | P08476 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 3624 |
| Molecular Weight (g/mol) | M.W. (Observed): 14 kDa, reducing conditions, 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer) |
| Name | ProDots™ Recombinant Human/Mouse/Rat Activin A |
| Quantity | 250 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2°C to 8°C as supplied. 1 month at 2°C to 8°C under sterile conditions after reconstitution. 3 months (at -20°C to -80°C under sterile conditions after reconstitution.) |
| Structural Form | Disulfide-linked homodimer |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.