Learn More
R&D Systems™ ProDots Recombinant Human TGF-beta 1
Description
Includes
Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human TGF-beta 1.
Order Info
Add ProDots proteins directly to the medium or reconstitute in the vial at 20μg/mL in 4mM HCl.
Specifications
Specifications
| Accession Number | P01137 |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | Predicted MW: 12.8kDa (monomer) Observed MW: 12kDa, reducing conditions, 24kDa, non-reducing conditions |
| Name | ProDots™ Recombinant Human TGF-beta 1 |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Ala279-Ser390 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Gene Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.