Learn More
Strem, An Ascensus Company CAS# 919778-41-9. 50mg. (R,R)-(+)-1,2-Bis(t-butylmethylphosphino)benzene (R,R)-BenzP*. MFCD18837015
SDP
Supplier:  Strem, An Ascensus Company 15016650MG
Encompass_Preferred
CAS# 919778-41-9. 50mg. (R,R)-(+)-1,2-Bis(t-butylmethylphosphino)benzene (R,R)-BenzP*. MFCD18837015. Molecular Weight: 282.34. Molecular Formula: C16H28P2. Color/form: white xtl. Strem# 15-0166. http://www.strem.com/catalog/v/15-0166/
Catalog No. 50-204-5233
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.