Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-Ts-DENEB(regR), TCI America™
SDP

Catalog No. T30781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30781G 1 g
T3078200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30781G Supplier TCI America Supplier No. T30781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1333981-84-2
Molecular Formula C31H33ClN2O3RuS
Molecular Weight (g/mol) 650.196
MDL Number MFCD20922902
InChI Key INKUCOHLIHBSDN-ZAMYOOMVSA-M
Synonym Chloro[(R,R)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulfonamide]ruthenium(II), Chloro[(R,R)-N-[2-(4-methylbenzyloxy)ethyl]-N′C-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II)
PubChem CID 121235224
IUPAC Name chlororuthenium(1+);[(1R,2R)-2-[2-[(4-methylphenyl)methoxy]ethylamino]-1,2-diphenylethyl]-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)COCCNC(C2=CC=CC=C2)C(C3=CC=CC=C3)[N-]S(=O)(=O)C4=CC=C(C=C4)C.Cl[Ru+]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 1 g
UN Number 3077
Formula Weight 650.20
Chemical Name or Material (R,R)-Ts-DENEB(regR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.