Learn More
CELLTREAT Scientific Products Multiple Cavity Reagent Reservoir, 8 Well, Trough Bottom, PP, Non-sterile, 25/CS
Supplier: CELLTREAT Scientific Products 229561
CELLTREAT Scientific Products Multiple Cavity Reagent Reservoir, 8 Well, Trough Bottom. Reagent Reservoirs are designed to assist in repetitive or robotic liquid transfer. Trough Bottom design promotes funneling of liquid to maximize reagent recovery by pipet tips. Low profile is ideal for small volume robotic tips. Multiple wells allow for dispensing of various reagents. Polypropylene offers a low binding surface to prevent samples from sticking to the side during elution, and is chemically inert for combinatorial chemistry applications. Storage possible at -80¿C. Autoclavable. Manufactured from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged Non-sterile, 5/bag, 25/case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.