Learn More
Abcam (R,S)-3,5-DHPG, Group I mGlu Receptor Agonist, 10 mM/mL
Description
- Molecular weight: 183.16 Da
- Purity: >98%
- Description: Selective group I mGlu receptor agonist
- Constituents: 100% (R,S)-3,5-DHPG
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Content And Storage | Store at -20°C. |
| Buffer | 100% (R,S)-3,5-DHPG |
| Shipping Condition | Ambient |
| Target Molecule | Group I mGlu Receptor |
| Formulation | 100% (R,S)-3,5-DHPG |
| Quantity | 10 mM/mL |
| For Use With (Application) | mGlu receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.