Learn More
Abcam (R,S)-AMPA, AMPA Agonist, Solid, >99% Purity, 10 mM/mL
Description
- Purity: >99%
- Molecular Weight: 186.17 Da
- Form: Solid
- Description: Prototypic agonist for AMPA receptors
- Storage: Store at ambient temperature
- Shipping: Ambient - can ship with ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | AMPA Receptors |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% (R,S)-AMPA |
| Form | Solid |
| Formulation | 100% (R,S)-AMPA |
| Quantity | 10 mM/mL |
| For Use With (Application) | AMPA receptor agonist |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.