Learn More
StemRD R SPONDIN-1 (HUMAN) 5MC
Supplier:  StemRD RSPO005
Encompass_Preferred
Proteins, Growth Factor Products, R SPONDIN-1 (HUMAN). A natural enhancer of the canonical WNT pathway. When used together with WNT proteins, enhance activation of the beta-catenin pathway. Made in human cells. Lyophilized powder
Catalog No. 50-752-7097
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.