Learn More
Medchemexpress LLC (R)-Zinc-3573 | 2089389-15-9 | 99.8% | 307.39 | C18H21N5 | 25 MG

Supplier: Medchemexpress LLC HY11806925MG
(R)-ZINC-3573 is a selective agonist of the Mas-related G protein-coupled receptor X2 (MRGPRX2) and is used as an in vitro research probe to study MRGPRX2-mediated signaling, mast cell degranulation, and mechanisms underlying pain and itch.
- Selective MRGPRX2 agonist for targeted receptor studies.
- Useful in vitro probe for mast cell degranulation and calcium signaling assays.
- Molecular formula C18H21N5; molecular weight 307.39 g/mol.
- High purity (≈99.8%) suitable for biochemical and pharmacological assays.
- Soluble in DMSO for standard assay preparations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.