Learn More
TARGETMOL CHEMICALS INC ZTZ240 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2923625MG
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ZTZ240 is a KCNQ2 channel activator used in the study of epilepsy. Purity 99.93%
Catalog No. 50-287-9441
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.