Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ R10000 Series Printer Ribbon

Catalog No. 19129405 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Green
Magenta
Red
White
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-129-405 Green
19-129-403 Black
19-129-404 Blue
19-129-407 Red
19-129-408 White
19-129-409 Yellow
19-129-406 Magenta
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-129-405 Supplier Brady™ Supplier No. M61R10010GN
Add to Cart
Add to Cart

Switch out your ribbon in seconds with these easy-to-load ribbon cartridges that slide into place in the always-correct direction.

  • Make it last – R10000 series ribbons are based on a resin formulation that produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • Make it fast – self-contained ribbon cartridges are easy to replace eliminating the hassle of printer calibration and ribbon threading
  • R10000 series ribbons are ideal for use with long-lasting indoor / outdoor materials such as Brady B-595

Specifications

Product Type Printer Ribbon
Color Green
For Use With (Equipment) BMP61 Labeling System, BradyPrinter M611
Material Resin
Print Technology Thermal Transfer
Length (English) 75 ft.
Width (English) 2 in.
Dimensions (L x W) 75 ft. x 2 in.
Alternate Part Number M61-R10010-GN
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.