Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ R18
GREENER_CHOICE

Catalog No. 21441
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-441 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-441 Supplier Tocris Bioscience™ Supplier No. 2144/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of 14.3.3 proteins

R18 is an antagonist of 14.3.3 proteins (KD ≈80 nM). Competitively inhibits 14.3.3-ligand interactions without requiring phosphorylation. Blocks the ability of 14.3.3 to bind to target proteins such as Raf-1, Bad, ASK1 and exoenzyme S.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Description R18
Molecular Weight (g/mol) 2309.69
Product Type R18
Quantity 1 mg
Sequence Pro-His-Cys-Val-Pro-Arg-Asp-Leu-Ser-Trp-Leu-Asp-Leu-Glu-Ala-Asn-Met-Cys-Leu-Pro

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.