Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ R3HCC1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101614 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101614 Supplier Invitrogen™ Supplier No. PA5101614
  • $587.00
    $489.65 / Each of 1

    Save $97.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GIDRP88.

R3HCC1L gene ontology (GO) annotation includes protein binding and exon-exon junction complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen R3HCC1L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene R3HCC1L
Gene Accession No. Q7Z5L2, Q8BJM3
Gene Alias 1700036B12Rik; AI450607; C10orf28; Coiled-coil domain-containing protein R3HCC1L; D19Ertd386e; GIDRP86; Gidrp88; growth inhibition and differentiation related protein 86; growth inhibition and differentiation-related protein 88; Growth inhibition and differentiation-related protein 88 homolog; PSORT; Putative mitochondrial space protein 32.1; R3H and coiled-coil domain-containing protein 1-like; R3H domain and coiled-coil containing 1 like; R3H domain and coiled-coil containing 1-like; R3hcc1l; RGD1307934
Gene Symbols R3HCC1L
Host Species Rabbit
Immunogen A synthesized peptide derived from human R3HCC1L(Accession Q7Z5L2), corresponding to amino acid residues K730-R780.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27291, 293953, 52013
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.