Learn More
Cytek Biosciences Inc cFluor® R668 Anti-Human IgG (4A11) - 25 Tests
Supplier:  Cytek Biosciences Inc R720390
Encompass
The 4A11 monoclonal antibody binds to the Fab fragment of human IgG. IgG is a membrane BCR that is produced when activated naïve B cells undergo class switch recombination of the Ig heavy chain constant region. This class-switching results in IgM and IgD being replaced by IgG as the activated B cell develops into memory and antibody secreting cells. On memory B cells, when IgG binds to its target antigen, it initiates signaling and activation that rapidly leads to proliferation and differentiation into antibody secreting cells. The antibody was conjugated to a fluorophore and purified by chromatography.
Catalog No. NC2888873
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.