Learn More
United States Biological Corporation 037167-PE Rabbit Anti-ITPRIPL1, NT (ITPRIPL1, KIAA1754L, Inositol 1,4,5-trisphosphate receptor-interacting protein-like 1) (PE)
Supplier:  United States Biological Corporation 037167PE200UL
Encompass_Preferred
ITPRIPL1 (inositol 1,4,5-triphosphate receptor-interacting protein-like 1), also known as KIAA1754L, is a 555 amino acid protein belonging to the ITPRIP family. ITPRIPL1 is a single-pass type I membrane protein expressed as two isoforms produced by alternative splicing events.
Catalog No. NC2947750
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.