Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579901
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579901 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579901 Supplier Invitrogen™ Supplier No. PA579901
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human Hela whole cell, human A549 whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue, mouse NIH/3T3 whole cell.

RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB10
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene RAB10
Gene Accession No. P35281, P61026, P61027
Gene Alias Ac1075; AW107754; GTP-binding protein RAB10; Rab10; RAB10, member RAS oncogene family; ras-related GTP-binding protein; ras-related protein rab10; ras-related protein Rab-10
Gene Symbols RAB10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human RAB10 (153-185aa AKANINIEKAFLTLAEDILRKTPVKEPNSENVD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10890, 19325, 50993
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.