Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118915 Supplier Invitrogen™ Supplier No. PA5118915
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB13.

RAB13 can participate in polarized transport, in the assembly and/or the activity of tight junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB13
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB13
Gene Accession No. P35286, P51153, Q9DD03
Gene Alias 0610007N03Rik; B230212B15Rik; Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; Rab13; RAB13, member RAS oncogene family; RAS-associated protein RAB13; ras-related protein Rab-13
Gene Symbols RAB13
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB13(Accession P51153), corresponding to amino acid residues E68-L118.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5872, 68328, 81756
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.