Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB22A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104069
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104069 Supplier Invitrogen™ Supplier No. PA5104069
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB22A.

The protein encoded by this gene is a member of the RAB family of small GTPases. The GTP-bound form of the encoded protein has been shown to interact with early-endosomal antigen 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB22A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB22A
Gene Accession No. P35285, Q9UL26
Gene Alias 3732413A17Rik; AI662177; AW319644; AW550514; E130120E14Rik; GTP-binding protein RAB22A; im:7136651; MGC16770; rab-14; Rab22; Rab-22; RAB22, member RAS oncogene family; Rab22a; Rab22a protein; RAB22A, member RAS oncogene family; RAS-related protein RAB-22A; zgc:101631
Gene Symbols RAB22A
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB22A(Accession Q9UL26), corresponding to amino acid residues F159-C194.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19334, 366265, 57403
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.