Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB30 Monoclonal Antibody (OTI3F4)
GREENER_CHOICE

Catalog No. PIMA526111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526111 Supplier Invitrogen™ Supplier No. MA526111
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB30 Monoclonal Antibody for Western Blot, ICC/IF

RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB30
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB30
Gene Accession No. Q15771, Q923S9
Gene Alias 5033421K01Rik; AI323892; EGK_06727; Rab30; RAB30, member RAS oncogene family; ras-related protein Rab-30; Rsb30
Gene Symbols RAB30
Host Species Mouse
Immunogen Full length human recombinant protein of RAB30 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27314, 308821, 476787, 75985
Target Species Human, Mouse, Rat, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.