Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB39B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104077 Supplier Invitrogen™ Supplier No. PA5104077
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB39B.

This gene encodes a member of the Rab family of proteins. Rab proteins are small GTPases that are involved in vesicular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB39B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB39B
Gene Accession No. Q8BHC1, Q96DA2
Gene Alias 6330580M05Rik; MRX72; Rab39b; RAB39B, member RAS oncogene family; ras-related protein Rab-39B; ras-related protein Rab-39B; LOW QUALITY PROTEIN: ras-related protein Rab-39B; WSMN
Gene Symbols RAB39B
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB39B(Accession Q96DA2), corresponding to amino acid residues W103-S153.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116442, 67790
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.