Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3A Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
SDP

Catalog No. 501682858
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2858 Supplier OriGene Supplier No. TA809529
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.

Specifications

Antigen RAB3A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RAB3A
Gene Accession No. P20336
Gene Alias RAB3A, member RAS oncogene family
Gene Symbols RAB3A
Host Species Mouse
Immunogen Full length human recombit protein of human RAB3A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5864
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.