Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1770 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1770 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1770 Supplier Invitrogen™ Supplier No. PA1770
  • $680.50
    $502.65 / Each of 1

    Save $177.85

    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-770 detects rab 3A from canine, hamster, human, mouse and rat tissues and cells. PA1-770 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects an ~21 kDa protein representing rab 3A from Hepa1 cell lysate. PA1-770 immunogen is a synthetic peptide corresponding to residues M(1) A S A T D S R Y G Q K E S S D Q N(18) C of human RAB3A. This sequence is completely conserved between human, mouse, and rat rab 3A. PA1-770 immunizing peptide (Cat. # PEP-098) is available for use in neutralization and control experiments. Recombinant human rab 3A (Cat # RP-770) protein can be purchased for control experiments in Western blot applications.

Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene RAB3A
Gene Accession No. P20336, P63011, P63012
Gene Alias C18A3.6; RAB3; Rab3a; RAB3A, member RAS oncogene family; RAS-associated protein RAB3A; ras-related protein Rab-3A; ras-related protein Rab-3A-like protein; Ras-related small GTP binding protein 3A
Gene Symbols RAB3A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) A S A T D S R Y G Q K E S S D Q N(18) C of human RAB3A.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100759024, 19339, 25531, 484826, 5864
Target Species Human, Mouse, Rat, Canine, Hamster
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.