Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rab3A+Rab3B+Rab3C+Rab3D Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501987107
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7107 Supplier Bioss Supplier No. BS6180R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has four known isoforms: 3A, 3B, 3C and 3D. These proteins are expressed in the highest concentration in the brain and endocrine tissues and are involved in regulated exocytosis of hormones and neurotransmitters. Specifically Rab3′s are thought to play a role in presynaptic vesicle trafficking and priming.

Specifications

Antigen Rab3A+Rab3B+Rab3C+Rab3D
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RAB3A
Gene Accession No. O95716, P20336, P20337, P35276, P62823, P62824, P63011, P63012, Q63941, Q63942, Q96E17, Q9CZT8
Gene Symbols RAB3A, RAB3B, RAB3C, RAB3D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Rab3A+Rab3B+Rab3C+Rab3D.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115827, 140665, 171058, 19339, 19340, 25531, 5864, 5865, 67295, 69908, 81755, 9545
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.