Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598106
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598106 Supplier Invitrogen™ Supplier No. PA598106
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB3B Polyclonal Antibody for Western Blot, IHC (P), ELISA

Protein transport. Probably involved in vesicular traffic. Target protein may play a role in secretory granule function and exocytosis in pancreatic acinar cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Rab3b
Gene Accession No. P20337, Q9CZT8
Gene Alias 2610528C18Rik; brain antigen RAB3B; Rab3b; RAB3B, member RAS oncogene family; ras-related protein Rab-3B
Gene Symbols Rab3b
Host Species Rabbit
Immunogen Recombinant Human Ras-related protein Rab-3B protein (1-219AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5865, 69908
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.