Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3GAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537026 Supplier Invitrogen™ Supplier No. PA537026
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 111 kDa. Purity is >95% by SDS-PAGE.

RAB3GAP1, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3GAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB3GAP1
Gene Accession No. P69735, Q15042, Q80UJ7
Gene Alias 1700003B17Rik; 4732493F09Rik; AL117896; KIAA0066; P130; Rab 3 GAP p130; Rab3 GAP p130; RAB3 GTPase activating protein catalytic subunit 1; RAB3 GTPase activating protein subunit 1; RAB3 GTPase activating protein subunit 1 (catalytic); RAB3 GTPase-activating protein 130 kDa subunit; Rab3 GTPase-activating protein catalytic subunit; RAB3GAP; Rab3-GAP; Rab3-GAP p130; Rab3gap1; RAB3GAP130; RGD1306487; WARBM1
Gene Symbols RAB3GAP1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 580-625 of human RAB3GAP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226407, 22930, 304759
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.