Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3IL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107011 Supplier Invitrogen™ Supplier No. PA5107011
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 100%; Rat: 100%.

Guanine nucleotide exchange factor (GEF) for Rab3A, a GTPase that regulates synaptic vesicle exocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3IL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene Rab3il1
Gene Accession No. Q8TBN0
Gene Alias 1200014K04Rik; AI115013; C76746; Grab; guanine nucleotide exchange factor for Rab3A; Guanine nucleotide exchange factor for Rab-3A; RAB3A interacting protein (rabin3)-like 1; RAB3A interacting protein like 1; Rab3ail1; rab-3A-interacting-like protein 1; rab3A-interacting-like protein 1; RAB3IL1; Rabin3-like 1
Gene Symbols Rab3il1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RAB3IL1 (aa 143-193).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5866
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.