Learn More
Invitrogen™ RAB3IP Monoclonal Antibody (OTI4A7)

Description
RAB3IP Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | RAB3IP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4A7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RAB3IP |
| Gene Accession No. | Q96QF0 |
| Gene Alias | B230311A06; gene trap PAT 12; Gtpat12; MGC137223 protein; RAB3A interacting protein; RAB3A interacting protein (rabin3); rab-3A-interacting protein; RAB3A-interacting protein; RAB3IP; rab3ip {ECO:0000250; rabin 3; Rabin3; rabin-3; RABIN8; SSX2 interacting protein; SSX2-interacting protein; UniProtKB:Q96QF0} |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.