Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rab4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674575 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-575 Supplier Invitrogen™ Supplier No. PA3912
  • $644.78
    $502.65 / Each of 1

    Save $142.13

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-912 detects Rab4 from human and mouse samples. This antibody does not detect other Rab isoforms (Rab1, Rab3, Rab5, or Rab6). PA3-912 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a ~24 kDa band representing Rab4 from transfected HeLa cells. The PA3-912 immunogen is purified recombinant human Rab4 protein.

Protein transport. Probably involved in vesicular traffic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene Rab4a
Gene Accession No. P05714, P20338, P56371
Gene Alias AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4
Gene Symbols Rab4a
Host Species Rabbit
Immunogen Purified, recombinant human Rab4 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19341, 25532, 479210, 5867
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.