Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551932 Supplier Invitrogen™ Supplier No. PA551932
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSLLFMETSA KTAMNVNEIF MAIAKKLPKN EPQNATGAPG RNRGVDLQEN NPASRSQCCS N Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. The transport of newly synthesized proteins from the endoplasmic reticulum to various stacks of the Golgi complex and to secretory vesicles involves at each stage the movement of carrier vesicles, a process that appears to involve Rab protein function. The possibility that Rab proteins might also direct the exocytosis from secretory vesicles to the plasma membrane is supported by the observation that in yeast, the SEC4 protein, which is 40% homologous to Rab proteins, is associated with secretory vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5C
Applications Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB5C
Gene Accession No. P51148
Gene Alias AI326010; L1880; RAB; RAB5B; RAB5B, member RAS oncogene family; RAB5C; Rab5c protein; RAB5C, member of RAS oncogene family; RAB5C, member RAS oncogene family; RAB5C, member RAS oncogene family isoform b; RAB5CL; RAB5L; RABL; Ras-related protein Rab-5C; small GTPase; wu:fb98b08; wu:fi16g01
Gene Symbols RAB5C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB5C. Recombinant protein control fragment (Product #RP-102038).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5878
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.